Order Entry
United States
ContactUsLinkComponent
Anti-SOX21 Mouse Monoclonal Antibody [clone: 2G10]
Anti-SOX21 Mouse Monoclonal Antibody [clone: 2G10]
Catalog # 103340-292
Supplier:  Novus Biologicals
Anti-SOX21 Mouse Monoclonal Antibody [clone: 2G10]
Catalog # 103340-292
Supplier:  Novus Biologicals
Supplier Number:  H00011166-M01

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    SOX21
  • Clonality:
    Monoclonal
  • Clone:
    2G10
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μg
  • Gene ID:
    11166
  • Antigen Synonyms:
    SOX25SOX-A|SRY (sex determining region Y)-box 21|SRY-box 21|transcription factor SOX-21|SOXA
  • Storage Buffer:
    PBS (pH 7.4)
  • Storage Temperature:
    Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
  • Immunogen:
    SOX21 (NP_009015, 224 a.a. ~ 266 a.a) partial recombinant protein with GST tag.HSHPSPGNPGYMIPCNCSAWPSPGLQPPLAYILLPGMGKPQL
  • Purification:
    IgG purified
  • Cat. No.:
    103340-292
  • Supplier no.:
    H00011166-M01

Specifications

About this item

The SOX21 Antibody (2G10) from Novus Biologicals is a mouse monoclonal antibody to SOX21. This antibody reacts with human. The SOX21 Antibody (2G10) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: SOX21
Clonality: Monoclonal
Clone: 2G10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human