Order Entry
United States
ContactUsLinkComponent
Anti-SOX11 Rabbit Polyclonal Antibody
Anti-SOX11 Rabbit Polyclonal Antibody
Catalog # 103270-030
Supplier:  Novus Biologicals
Anti-SOX11 Rabbit Polyclonal Antibody
Catalog # 103270-030
Supplier:  Novus Biologicals
Supplier Number:  NBP1-85823

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    SOX11
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Gene ID:
    6664
  • Antigen Synonyms:
    SRY (sex-determining region Y)-box 11|SRY (sex determining region Y)-box 11|transcription factor SOX-11|SRY-related HMG-box gene 11
  • Storage Buffer:
    PBS (pH 7.2) and 40% Glycerol
  • Storage Temperature:
    Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
  • Immunogen:
    This antibody was developed against Recombinant Protein corresponding to amino acids:FMVWSKIERRKIMEQSPDMHNAEISKRLGKRWKMLKDSEKIPFIREAERLRLKHMADYPDYKYRPRKKPKMDPSAKPSASQSPEKSAAGGGGGSAGGGAGGAKTSKGSSKK
  • Purification:
    Immunogen affinity purified
  • Cat. No.:
    103270-030
  • Supplier no.:
    NBP1-85823

Specifications

About this item

The SOX11 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SOX11. This antibody reacts with human. The SOX11 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SOX11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human