Anti-SNX33 Rabbit Polyclonal Antibody
Catalog # 103348-018
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:SNX33
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:257364
- Antigen Synonyms:SNX30SH3 and PX domain-containing protein 3|SH3PXD3CSH3PX3|sorting nexin-33|SH3 and PX domain containing 3|MGC32065|sorting nexin 33
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:SH3PX3 (NP_695003.1, 1 a.a. ~ 574 a.a) full-length human protein.MALKGRALYDFHSENKEEISIQQDEDLVIFSETSLDGWLQGQNSRGETGLFPASYVEIVRSGISTNHADYSSSPAGSPGAQVSLYNSPSVASPARSGGGSGFLSNQGSFEEDDDDDWDDWDDGCTVVEEPRAGGLGTNGHPPLNLSYPGAYPSQHMAFRPKPPLERQDSLASAKRGSVVGRNLNRFSCFVRSGVEAFILGDVPMMAKIAETYSIEMGPRGPQWKANPHPFACSVEDPTKQTKFKGIKSYISYKLT
- Purification:Protein A purified
- Cat. No.:103348-018
- Supplier no.:H00257364-D01P
Specifications
About this item
The SNX33 Antibody from Novus Biologicals is a rabbit polyclonal antibody to SNX33. This antibody reacts with human. The SNX33 Antibody has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: SNX33
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human