Order Entry
United States
ContactUsLinkComponent
Anti-SNX1 Mouse Monoclonal Antibody [clone: 1E3.]
Anti-SNX1 Mouse Monoclonal Antibody [clone: 1E3.]
Catalog # 103310-492
Supplier:  Novus Biologicals
Anti-SNX1 Mouse Monoclonal Antibody [clone: 1E3.]
Catalog # 103310-492
Supplier:  Novus Biologicals
Supplier Number:  H00006642-M02

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    SNX1
  • Clonality:
    Monoclonal
  • Clone:
    1E3.
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μg
  • Gene ID:
    6642
  • Antigen Synonyms:
    Vps5|MGC8664|HsT17379|sorting nexin 1|SNX1Asorting nexin 1A|sorting nexin-1
  • Storage Buffer:
    PBS (pH 7.4)
  • Storage Temperature:
    Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
  • Immunogen:
    SNX1 (NP_003090, 166 a.a. - 275 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. KVTTQTSLPLFRSKQFAVKRRFSDFLGLYEKLSEKHSQNGFIVPPPPEKSLIGMTKVKVGKEDSSSAEFLEKRRAALERYLQRIVNHPTMLQDPDVREFLEKEELPRAVG
  • Purification:
    IgG purified
  • Cat. No.:
    103310-492
  • Supplier no.:
    H00006642-M02

Specifications

About this item

The SNX1 Antibody (1E3.) from Novus Biologicals is a mouse monoclonal antibody to SNX1. This antibody reacts with human. The SNX1 Antibody (1E3.) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: SNX1
Clonality: Monoclonal
Clone: 1E3.
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human