Anti-Six3 Mouse Monoclonal Antibody [clone: 2B9]
Catalog # 103310-186
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:Six3
- Clonality:Monoclonal
- Clone:2B9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:6496
- Antigen Synonyms:HPE2|SIX homeobox 3|homeobox protein SIX3|holoprosencephaly 2|Sine oculis homeobox homolog 3|sine oculis homeobox homolog 3 (Drosophila)|alobar or semilobar
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:SIX3 (NP_005404 273 a.a. - 332 a.a.) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. QAIGPSGMRSLAEPGCPTHGSAESPSTAASPTTSVSSLTERADTGTSILSVTSSDSECDV
- Purification:IgG purified
- Cat. No.:103310-186
- Supplier no.:H00006496-M08
Specifications
About this item
The Six3 Antibody (2B9) from Novus Biologicals is a mouse monoclonal antibody to Six3. This antibody reacts with human. The Six3 Antibody (2B9) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: Six3
Clonality: Monoclonal
Clone: 2B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human