Anti-SGSH Mouse Polyclonal Antibody
Catalog # 103310-090
Supplier: Avantor
Specifications
- Antibody Type:Primary
- Antigen Name:Sulfamidase
- Antigen Symbol:SGSH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Gene ID:6448
- Antigen Synonyms:heparan sulfate sulfatase|Sulphamidase|SFMDEC 3.10.1.1|Sulfoglucosamine sulfamidase|N-sulfoglucosamine sulfohydrolase|HSSN-sulphoglucosamine sulphohydrolase|MPS3Asulphamidase
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:SGSH (NP_000190.1, 1 a.a. - 502 a.a.) full-length human protein. MSCPVPACCALLLVLGLCRARPRNALLLLADDGGFESGAYNNSAIATPHLDALARRSLLFRNAFTSVSSCSPSRASLLTGLPQHQNGMYGLHQDVHHFNSFDKVRSLPLLLSQAGVRTGIIGKKHVGPETVYPFDFAYTEENGSVLQVGRNITRIKLLVRKFLQTQDDRPFFLYVAFHDPHRCGHSQPQYGTFCEKFGNGESGMGRIPDWTPQAYDPLDVLVPYFVPNTPAARADLAAQYTTVGRMDQGVGLVLQELRDAGVLNDTLVIFTSDNGIPFPSGRTNLYWPGTAEPLLVSSPEHPKRWGQVSEAYVSLLDLTPTILDWFSIPYPSYAIFGSKTIHLTGRSLLPALEAEPLWATVFGSQSHHEVTMSYPMRSVQHRHFRLVHNLNFKMPFPIDQDFYVSPTFQDLLNRTTAGQPTGWYKDLRHYYYRARWELYDRSRDPHETQNLATDPRFAQLLEMLRDQLAKWQWETHDPWVCAPDGVLEEKLSPQCQPLHNEL
- Purification:Protein A purified
- Cat. No.:103310-090
- Supplier no.:H00006448-B01P
Specifications
About this item
The Sulfamidase / SGSH Antibody from Novus Biologicals is a mouse polyclonal antibody to Sulfamidase / SGSH. This antibody reacts with human. The Sulfamidase / SGSH Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: SGSH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human