Anti-SGK2 Mouse Monoclonal Antibody [clone: 2F6]
Catalog # 103338-170
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:SGK2
- Clonality:Monoclonal
- Clone:2F6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:10110
- Antigen Synonyms:serum/glucocorticoid regulated kinase 2|dJ138B7.2|EC 2.7.11.1|EC 2.7.11|serine/threonine-protein kinase Sgk2|Serum/glucocorticoid-regulated kinase 2|H-SGK2
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:SGK2 (AAH65511, 293 a.a. ~ 367 a.a) partial recombinant protein with GST tag.SPINWDDLYHKRLTPPFNPNVTGPADLKHFDPEFTQEAVSKSIGCTPDTVASSSGASSAFLGFSYAPEDDDILDC
- Purification:IgG purified
- Cat. No.:103338-170
- Supplier no.:H00010110-M07
Specifications
About this item
The SGK2 Antibody (2F6) from Novus Biologicals is a mouse monoclonal antibody to SGK2. This antibody reacts with human. The SGK2 Antibody (2F6) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: SGK2
Clonality: Monoclonal
Clone: 2F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG Mix Kappa
Reactivity: Human