Anti-SERBP1 Mouse Monoclonal Antibody [clone: 1D2-2E9]
Catalog # 103342-132
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:SERBP1
- Clonality:Monoclonal
- Clone:1D2-2E9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:26135
- Antigen Synonyms:CGI-55|PAI-RBP1DKFZp564M2423|SERPINE1 mRNA binding protein 1|HABP4L|CHD3IP|PAI1 RNA-binding protein 1|plasminogen activator inhibitor 1 RNA binding protein|plasminogen activator inhibitor 1 RNA-binding protein|chromodomain helicase DNA binding protein 3 interacting protein|PAI-1 mRNA binding protein|SERPINE1 mRNA-binding protein 1|PAIRBP1FLJ90489|DKFZP564M2423
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:PAI-RBP1 (AAH02488, 1 a.a. ~ 403 a.a) full length recombinant protein with GST tag.MPGHLQEGFGCVVTNRFDQLFDDESDPFEVLKAAENKKKEAGGGGVGGPGAKSAAQAAAQTNSNAAGKQLRKESQKDRKNPLPPSVGVVDKKEETQPPVALKKEGIRRVGRRPDQQLQGEGKIIDRRPERRPPRERRFEKPLEEKGEGGEFSVDRPIIDRPIRGRGGLGRGRGGRGRGMGRGDGFDSRGKREFDRHSGSDRSGLKHEDKRGGSGSHNWGTVKDELTESPKYIQKQISYNYSDLDQSNVTEETPEG
- Purification:IgG purified
- Cat. No.:103342-132
- Supplier no.:H00026135-M01
Specifications
About this item
The SERBP1 Antibody (1D2-2E9) from Novus Biologicals is a mouse monoclonal antibody to SERBP1. This antibody reacts with human. The SERBP1 Antibody (1D2-2E9) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: SERBP1
Clonality: Monoclonal
Clone: 1D2-2E9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human