Anti-Salivary Amylase Alpha Mouse Monoclonal Antibody [clone: 3C9]
Catalog # 103321-796
Supplier: Avantor
Specifications
- Antibody Type:Primary
- Antigen Name:Salivary Amylase Alpha
- Antigen Symbol:Salivary Amylase Alpha
- Clonality:Monoclonal
- Clone:3C9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Size:100 μg
- Gene ID:276
- Antigen Synonyms:salivary|AMY1B|AMY1|salivary amylase alpha 1A|alpha 1A|AMY1C|glycogenase|alpha 1A (salivary)|alpha-1A|1,4-alpha-D-glucan glucanohydrolase 1|amylase|Salivary alpha-amylase|alpha-amylase 1|EC 3.2.1.1
- Storage Buffer:PBS (pH 7.2)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:Native purified human AMY1A.
- Purification:IgG purified
- Cat. No.:103321-796
- Supplier no.:H00000276-M11
Specifications
About this item
The Salivary Amylase Alpha Antibody (3C9) from Novus Biologicals is a mouse monoclonal antibody to Salivary Amylase Alpha. This antibody reacts with human. The Salivary Amylase Alpha Antibody (3C9) has been validated for the following applications: ELISA.
Type: Primary
Antigen: Salivary Amylase Alpha
Clonality: Monoclonal
Clone: 3C9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human