Anti-RPS20 Mouse Monoclonal Antibody [clone: 1G12]
103309-588
- Antibody Type:Primary
- Antigen Symbol:RPS20
- Clonality:Monoclonal
- Clone:1G12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Size:100 μg
- Gene ID:6224
- Antigen Synonyms:40S ribosomal protein S20|ribosomal protein S20|MGC102930|FLJ27451
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:RPS20 (AAH07507, 1 a.a. - 119 a.a.) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MAFKDTGKTPVEPEVAIHRIRITLTSRNVKSLEKVCADLIRGAKEKNLKVKGPVRMPTKTLRITTRKTPCGEGSKTWDRFQMRIHKRLIDLHSPSEIVKQITSISIEPGVEVEVTIADA
- Purification:IgG purified
- Cat. No.:103309-588
- Supplier no.:H00006224-M03
The RPS20 Antibody (1G12) from Novus Biologicals is a mouse monoclonal antibody to RPS20. This antibody reacts with human. The RPS20 Antibody (1G12) has been validated for the following applications: ELISA.
Type: Primary
Antigen: RPS20
Clonality: Monoclonal
Clone: 1G12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human