To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:RAMP1
- Clonality:Monoclonal
- Clone:1F1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Size:100 μg
- Environmentally Preferable:
- Gene ID:10267
- Antigen Synonyms:receptor (G protein-coupled) activity modifying protein 1|receptor (calcitonin) activity modifying protein 1|calcitonin receptor-like receptor activity modifying protein 1|CRLR activity-modifying protein 1|receptor activity-modifying protein 1|receptor activity modifying protein 1|Calcitonin-receptor-like receptor activity-modifying protein 1
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:RAMP1 (NP_005846, 27 a.a. ~ 118 a.a) partial recombinant protein with GST tag.CQEANYGALLRELCLTQFQVDMEAVGETLWCDWGRTIRSYRELADCTWHMAEKLGCFWPNAEVDRFFLAVHGRYFRSCPISGRAVRDPPGS
- Purification:IgG purified
- Cat. No.:103338-558
- Supplier no.:H00010267-M01
The RAMP1 Antibody (1F1) from Novus Biologicals is a mouse monoclonal antibody to RAMP1. This antibody reacts with human. The RAMP1 Antibody (1F1) has been validated for the following applications: ELISA.
Type: Primary
Antigen: RAMP1
Clonality: Monoclonal
Clone: 1F1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human