Anti-Rad21 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: CM110-2C10]
Catalog # 103357-956
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:Rad21
- Clonality:Monoclonal
- Clone:CM110-2C10
- Conjugation:Alexa Fluor® 488
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:5885
- Antigen Synonyms:KIAA0078RAD21 (S. pombe) homolog|MCD1|RAD21 homolog (S. pombe)|Nuclear matrix protein 1|HR21|NXP-1|hHR21FLJ25655|NXP1HRAD21|protein involved in DNA double-strand break repair|double-strand-break repair protein rad21 homolog|SCC1FLJ40596|SCC1 homolog
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:An N-terminal fusion protein of the C-terminal fragment of Scc1 subunit human cohesin (amino acids 544-631 of hRad21).
- Purification:Protein G purified
- Cat. No.:103357-956
Specifications
About this item
The Rad21 Antibody (CM110-2C10) [Alexa Fluor« 488] from Novus Biologicals is a mouse monoclonal antibody to Rad21. This antibody reacts with human, mouse. The Rad21 Antibody (CM110-2C10) [Alexa Fluor« 488] has been validated for the following applications: Western Blot.
Type: Primary
Antigen: Rad21
Clonality: Monoclonal
Clone: CM110-2C10
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse