Anti-RAD18 Mouse Monoclonal Antibody (DyLight 488) [clone: 79B1048.1]
Catalog # 103359-660
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:RAD18
- Clonality:Monoclonal
- Clone:79B1048.1
- Conjugation:DyLight® 488
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:56852
- Antigen Synonyms:S. cerevisiae|EC 6.3.2.-|homolog|hHR18|postreplication repair protein hRAD18p|RNF73E3 ubiquitin-protein ligase RAD18|hRAD18|Postreplication repair protein RAD18|RAD18|RAD18 homolog (S. cerevisiae)|RING finger protein 73
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:The antibody was raised against a synthetic peptide corresponding to the amino acids 402-414 of human Rad18.
- Purification:Protein G purified
- Cat. No.:103359-660
Specifications
About this item
The RAD18 Antibody (79B1048.1) [DyLight 488] from Novus Biologicals is a mouse monoclonal antibody to RAD18. This antibody reacts with human. The RAD18 Antibody (79B1048.1) [DyLight 488] has been validated for the following applications: Western Blot.
Type: Primary
Antigen: RAD18
Clonality: Monoclonal
Clone: 79B1048.1
Conjugation: DyLight 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human