Anti-RAB40A Mouse Polyclonal Antibody
Catalog # 103347-046
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:RAB40A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Gene ID:142684
- Antigen Synonyms:RAR2A|MGC142061|SOCS box-containing protein RAR2A|Protein Rar-2|SOCS box containing protein RAR2A|RAB40A|ras-related protein Rab-40A|member RAS oncogene family|RAR2|Rar-2
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:RAB40A (NP_543155.2, 1 a.a. ~ 277 a.a) full-length human protein.MSAPGSPDQAYDFLLKFLLVGDRDVGKSEILESLQDGAAESPYSHLGGIDYKTTTILLDGQRVKLKLWDTSGQGRFCTIFRSYSRGAQGVILVYDIANRWSFEGMDRWIKKIEEHAPGVPKILVGNRLHLAFKRQVPREQAQAYAERLGVTFFEVSPLCNFNIIESFTELARIVLLRHRMNWLGRPSKVLSLQDLCCRTIVSCTPVHLVDKLPLPSTLRSHLKSFSMAKGLNARMMRGLSYSLTTSSTHKSSLCK
- Purification:Protein A purified
- Cat. No.:103347-046
- Supplier no.:H00142684-B01P
Specifications
About this item
The RAB40A Antibody from Novus Biologicals is a mouse polyclonal antibody to RAB40A. This antibody reacts with human. The RAB40A Antibody has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: RAB40A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human