Anti-PTDSS1 Mouse Polyclonal Antibody
103337-616
- Antibody Type:Primary
- Antigen Symbol:PTDSS1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Gene ID:9791
- Antigen Synonyms:PSSAPSS-1|phosphatidylserine synthase 1|PtdSer synthase 1|KIAA0024Serine-exchange enzyme I|PSS1EC 2.7.8.-
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:PTDSS1 (NP_055569.1, 1 a.a. ~ 473 a.a) full-length human protein.MASCVGSRTLSKDDVNYKMHFRMINEQQVEDITIDFFYRPHTITLLSFTIVSLMYFAFTRDDSVPEDNIWRGILSVIFFFLIISVLAFPNGPFTRPHPALWRMVFGLSVLYFLFLVFLLFLNFEQVKSLMYWLDPNLRYATREADVMEYAVNCHVITWERIISHFDIFAFGHFWGWAMKALLIRSYGLCWTISITWELTELFFMHLLPNFAECWWDQVILDILLCNGGGIWLGMVVCRFLEMRTYHWASFKDIHT
- Purification:Protein A purified
- Cat. No.:103337-616
- Supplier no.:H00009791-B01P
The PTDSS1 Antibody from Novus Biologicals is a mouse polyclonal antibody to PTDSS1. This antibody reacts with human. The PTDSS1 Antibody has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: PTDSS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human