Anti-PSRC2 Mouse Monoclonal Antibody [clone: 3A3]
Catalog # 103347-652
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:PSRC2
- Clonality:Monoclonal
- Clone:3A3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:196441
- Antigen Synonyms:zinc finger C3H1 domain-containing protein|MGC23401|PSRC2|coiled-coil domain containing 131|proline/serine-rich coiled-coil 2|Proline/serine-rich coiled-coil protein 2|zinc finger|MGC90200|C3H1-type containing|DKFZp686A0722|KIAA0546CCDC131|Coiled-coil domain-containing protein 131
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:CCDC131 (AAH15679, 1 a.a. ~ 359 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.MATADTPAPASSGLSPKEEGELEDGEISDDDNNSQIRSRSSSSSSGGGLLPYPRRRPPHSARGGGSGGGGGSSSSSSSSQQQLRNFSRSRHASERGHLRGPSSYRPKEPFRSHPPSVRMPSSSLSESSPRPSFWERSHLALDRFRFRGRPYRGGSRWSRGRGVGERGGKPGCRPPLGGGAGSGFSSSQSWREPSPPRKSSKSFGRSPSRKQNYSSKNENCVEETFEDLLLKYKQIQLELECINKDEKLALSSKEE
- Purification:IgG purified
- Cat. No.:103347-652
- Supplier no.:H00196441-M07
Specifications
About this item
The PSRC2 Antibody (3A3) from Novus Biologicals is a mouse monoclonal antibody to PSRC2. This antibody reacts with human. The PSRC2 Antibody (3A3) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: PSRC2
Clonality: Monoclonal
Clone: 3A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human