Anti-PRPF3 Rabbit Polyclonal Antibody
Catalog # 103259-016
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:PRPF3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:9129
- Antigen Synonyms:PRP3 pre-mRNA processing factor 3 homolog (S. cerevisiae)|U4/U6 snRNP 90 kDa protein|PRP3|Prp3p|PRP3 pre-mRNA processing factor 3 homolog (yeast)|RP18|Pre-mRNA-splicing factor 3|hPrp3|HPRP3retinitis pigmentosa 18 (autosomal dominant)|U4/U6 small nuclear ribonucleoprotein Prp3|U4/U6-associated RNA splicing factor|HPRP3P
- Storage Buffer:PBS (pH 7.2) and 40% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed against Recombinant Protein corresponding to amino acids:WWDSYIIPNGFDLTEENPKREDYFGITNLVEHPAQLNPPVDNDTPVTLGVYLTKKEQKKLRRQTRREAQKELQEKVRLGLMPPPEPKVRISNLMRVLGTEAVQDPTK
- Purification:Immunogen affinity purified
- Cat. No.:103259-016
- Supplier no.:NBP1-83219
Specifications
About this item
The PRPF3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PRPF3. This antibody reacts with human. The PRPF3 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: PRPF3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human