Anti-PRAT Rabbit Polyclonal Antibody
103307-858
: Avantor
- Antibody Type:Primary
- Antigen Name:Phosphoribosyl Pyrophosphate Amidotransferase
- Antigen Symbol:PRAT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:5471
- Antigen Synonyms:EC 2.4.2.14|GPATamidophosphoribosyltransferase|Glutamine phosphoribosylpyrophosphate amidotransferase|glutamine phosphoribosylpyrophosphatate amidotransferase|ATASE|PRAT|phosphoribosyl pyrophosphate amidotransferase|glutamine PRPP amidotransferase
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:PPAT (NP_002694.3, 1 a.a. - 517 a.a.) full-length human protein. MELEELGIREECGVFGCIASGEWPTQLDVPHVITLGLVGLQHRGQESAGIVTSDGSSVPTFKSHKGMGLVNHVFTEDNLKKLYVSNLGIGHTRYATTGKCELENCQPFVVETLHGKIAVAHNGELVNAARLRKKLLRHGIGLSTSSDSEMITQLLAYTPPQEQDDTPDWVARIKNLMKEAPTAYSLLIMHRDVIYAVRDPYGNRPLCIGRLIPVSDINDKEKKTSETEGWVVSSESCSFLSIGARYYREVLPGEIVEISRHNVQTLDIISRSEGNPVAFCIFEYVYFARPDSMFEDQMVYTVRYRCGQQLAIEAPVDADLVSTVPESATPAALAYAGKCGLPYVEVLCKNRYVGRTFIQPNMRLRQLGVAKKFGVLSDNFKGKRIVLVDDSIVRGNTISPIIKLLKESGAKEVHIRVASPPIKYPCFMGINIPTKEELIANKPEFDHLAEYLGANSVVYLSVEGLVSSVQEGIKFKKQKEKKHDIMIQENGNGLECFEKSGHCTACLTGKYPVELEW
- Purification:Protein G purified
- Cat. No.:103307-858
- Supplier no.:H00005471-D01P
The Phosphoribosyl Pyrophosphate Amidotransferase Antibody from Novus Biologicals is a rabbit polyclonal antibody to Phosphoribosyl Pyrophosphate Amidotransferase. This antibody reacts with human. The Phosphoribosyl Pyrophosphate Amidotransferase Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: PRAT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human