Anti-POP7 Mouse Monoclonal Antibody [clone: 3F12]
Catalog # 103338-510
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:POP7
- Clonality:Monoclonal
- Clone:3F12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 μg
- Gene ID:10248
- Antigen Synonyms:0610037N12Rik|EC 3.1.26.5|Ribonucleases P/MRP protein subunit POP7 homolog|ribonuclease P protein subunit p20|processing of precursor 7|hPOP7|RNaseP protein p20|RPP20S. cerevisiae) homolog|ribonuclease P/MRP subunit (S. cerevisiae)|ribonuclease P subunit|ribonuclease P subunit (S. cerevisiae)
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:POP7 (NP_005828, 1 a.a. ~ 81 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.MAENREPRGAVEAELDPVEYTLRKRLPSRLPRRPNDIYVNMKTDFKAQLARCQKLLDGGARGQNACSEIYIHGLGLAINH
- Purification:Protein A purified
- Cat. No.:103338-510
- Supplier no.:H00010248-M13
Specifications
About this item
The POP7 Antibody (3F12) from Novus Biologicals is a mouse monoclonal antibody to POP7. This antibody reacts with human. The POP7 Antibody (3F12) has been validated for the following applications: ELISA.
Type: Primary
Antigen: POP7
Clonality: Monoclonal
Clone: 3F12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human