Anti-POLR3D Mouse Polyclonal Antibody
Catalog # 103322-736
Supplier: Avantor
Specifications
- Antibody Type:Primary
- Antigen Symbol:POLR3D
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Gene ID:661
- Antigen Synonyms:BN51|44kDa|RNA polymerase III subunit C4|RNA polymerase III 47 kDa subunit|RPC53 homolog|polymerase (RNA) III (DNA directed) polypeptide D|tsBN51|TSBN51RPC53|temperature sensitive complementation|DNA-directed RNA polymerase III subunit D|RPC4|BN51TDNA-directed RNA polymerase III 47 kDa polypeptide|Protein BN51|BN51 (BHK21) temperature sensitivity complementing|cell cycle specific|DNA-directed RNA polymerase III subunit RPC4
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:POLR3D (NP_001713.2, 1 a.a. - 398 a.a.) full-length human protein. MSEGNAAGEPSTPGGPRPLLTGARGLIGRRPAPPLTPGRLPSIRSRDLTLGGVKKKTFTPNIISRKIKEEPKEEVTVKKEKRERDRDRQREGHGRGRGRPEVIQSHSIFEQGPAEMMKKKGNWDKTVDVSDMGPSHIINIKKEKRETDEETKQILRMLEKDDFLDDPGLRNDTRNMPVQLPLAHSGWLFKEENDEPDVKPWLAGPKEEDMEVDIPAVKVKEEPRDEEEEAKMKAPPKAARKTPGLPKDVSVAELLRELSLTKEEELLFLQLPDTLPGQPPTQDIKPIKTEVQGEDGQVVLIKQEKDREAKLAENACTLADLTEGQVGKLLIRKSGRVQLLLGKVTLDVTMGTACSFLQELVSVGLGDSRTGEMTVLGHVKHKLVCSPDFESLLDHKHR
- Purification:Protein A purified
- Cat. No.:103322-736
- Supplier no.:H00000661-B01P
Specifications
About this item
The POLR3D Antibody from Novus Biologicals is a mouse polyclonal antibody to POLR3D. This antibody reacts with human. The POLR3D Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: POLR3D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human