Anti-pIgR Rabbit Polyclonal Antibody
Catalog # 103272-528
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:pIgR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:5284
- Antigen Synonyms:polymeric immunoglobulin receptor|MGC125362|hepatocellular carcinoma associated protein TB6|MGC125361|Hepatocellular carcinoma-associated protein TB6|Poly-Ig receptor|FLJ22667|PIgR
- Storage Buffer:PBS (pH 7.2) and 40% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed against Recombinant Protein corresponding to amino acids:LLFSVVINQLRLSDAGQYLCQAGDDSNSNKKNADLQVLKPEPELVYEDLRGSVTFHCALGPEVANVAKFLCRQSSGENCDVVVNTLGKRAPAFEGRILLNPQDKDGSFSVVITGLRKEDAGRYLCGAHS
- Purification:Immunogen affinity purified
- Cat. No.:103272-528
- Supplier no.:NBP1-86095
Specifications
About this item
The pIgR Antibody from Novus Biologicals is a rabbit polyclonal antibody to pIgR. This antibody reacts with human. The pIgR Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: pIgR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human