Order Entry
United States
ContactUsLinkComponent
Anti-PGM3 Mouse Monoclonal Antibody [clone: 1E2-1B12]
Anti-PGM3 Mouse Monoclonal Antibody [clone: 1E2-1B12]
Catalog # 103332-698
Supplier:  Novus Biologicals
Anti-PGM3 Mouse Monoclonal Antibody [clone: 1E2-1B12]
Catalog # 103332-698
Supplier:  Novus Biologicals
Supplier Number:  H00005238-M01

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    PGM3
  • Clonality:
    Monoclonal
  • Clone:
    1E2-1B12
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μg
  • Gene ID:
    5238
  • Antigen Synonyms:
    Phosphoglucomutase-3|phosphoacetylglucosamine mutase|N-acetylglucosamine-phosphate mutase|FLJ13623|PGM 3|EC 5.4.2.3|AGM1PAGMAcetylglucosamine phosphomutase|FLJ11614|DKFZP434B187|N-acetylglucosamine-phosphate mutase 1|phosphoglucomutase 3|EC 5.4.2
  • Storage Buffer:
    PBS (pH 7.4)
  • Storage Temperature:
    Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
  • Immunogen:
    PGM3 (AAH01258, 1 a.a. - 542 a.a.) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MDLGAITKYSALHAKPNGLILQYGTAGFRTKAEHLDHVMFRMGLLAVLRSKQTKSTIGVMVTASHNPEEDNGVKLVDPLGEMLAPSWEEHATCLANAEEQDMQRVLIDISEKEAVNLQQDAFVVIGRDTRPSSEKLSQSVIDGVTVLGGQFHDYGLLTTPQLHYMVYCRNTGGRYGKATIEGYYQKLSKAFVELTKQASCSGDEYRSLKVDCANGIGALKLREMEHYFSQGLSVQLFNDGSKGKLNHLCGADFVKSHQKPPQGMEIKSNERCCSFDGDADRIVYYYHDADGHFHLIDGDKIATLISSFLKELLVEIGESLNIGVVQTAYANGSSTRYLEEVMKVPVYCTKTGVKHLHHKAQEFDIGVYFEANGHGTALFSTAVEMKIKQSAEQLEDKKRKAAKMLENIIDLFNQAAGDAISDMLVIEAILALKGLTVQQWDALYTDLPNRQLKVQVADRRVISTTNAERQAVTPPGLQEAINDLVKKYKLSRAFVRPSGTEDVVRVYAEADSQESADHLAHEVSLAVFQLAGGIGERPQPGF
  • Tested Applications:
    RNA Inhibition
  • Purification:
    IgG purified
  • Cat. No.:
    103332-698
  • Supplier no.:
    H00005238-M01

Specifications

About this item

The PGM3 Antibody (1E2-1B12) from Novus Biologicals is a mouse monoclonal antibody to PGM3. This antibody reacts with human. The PGM3 Antibody (1E2-1B12) has been validated for the following applications: Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry-Paraffin, RNA Inhibition.

Type: Primary
Antigen: PGM3
Clonality: Monoclonal
Clone: 1E2-1B12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human