Anti-PELO Rabbit Polyclonal Antibody
103272-272
- Antibody Type:Primary
- Antigen Symbol:PELO
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 μl
- Gene ID:53918
- Antigen Synonyms:pelota homolog (Drosophila)|protein pelota homolog|PRO1770|pelota (Drosophila) homolog|EC 3.1
- Storage Buffer:PBS (pH 7.2) and 40% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed against Recombinant Protein corresponding to amino acids:VTLVPEEPEDMWHTYNLVQVGDSLRASTIRKVQTESSTGSVGSNRVRTTLTLCVEAIDFDSQACQLRVKGTNIQENEYVKMGAYHTIELEPNRQFTLAKKQWDSVVLERIEQACDPAWSADVAAVVMQE
- Purification:Immunogen affinity purified
- Cat. No.:103272-272
- Supplier no.:NBP1-85923
The PELO Antibody from Novus Biologicals is a rabbit polyclonal antibody to PELO. This antibody reacts with human. The PELO Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: PELO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human