Anti-PCYT2 Rabbit Polyclonal Antibody
103308-844
- Antibody Type:Primary
- Antigen Symbol:PCYT2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:5833
- Antigen Synonyms:phosphate cytidylyltransferase 2|EC 2.7.7|CTP:phosphoethanolamine cytidylyltransferase|ET|EC 2.7.7.14|ethanolamine-phosphate cytidylyltransferase|ethanolamine|Phosphorylethanolamine transferase
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:PCYT2 (NP_002852.1, 1 a.a. - 389 a.a.) full-length human protein. MIRNGRGAAGGAEQPGPGGRRAVRVWCDGCYDMVHYGHSNQLRQARAMGDYLIVGVHTDEEIAKHKGPPVFTQEERYKMVQAIKWVDEVVPAAPYVTTLETLDKYNCDFCVHGNDITLTVDGRDTYEEVKQAGRYRECKRTQGVSTTDLVGRMLLVTKAHHSSQEMSSEYREYADSFGKCPGGRNPWTGVSQFLQTSQKIIQFASGKEPQPGETVIYVAGAFDLFHIGHVDFLEKVHRLAERPYIIAGLHFDQEVNHYKGKNYPIMNLHERTLSVLACRYVSEVVIGAPYAVTAELLSHFKVDLVCHGKTEIIPDRDGSDPYQEPKRRGIFRQIDSGSNLTTDLIVQRIITNRLEYEARNQKKEAKELAFLEAARQQAAQPLGERDGDF
- Purification:Protein G purified
- Cat. No.:103308-844
- Supplier no.:H00005833-D01P
The PCYT2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PCYT2. This antibody reacts with human. The PCYT2 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: PCYT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human