To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:PASK
- Clonality:Monoclonal
- Clone:6D10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:23178
- Antigen Synonyms:PAS-serine/threonine kinase|STK37|hPASK|PAS domain containing serine/threonine kinase|DKFZp434O051|EC 2.7.11.1|PASKINPAS domain-containing serine/threonine-protein kinase|PAS-kinase|KIAA0135DKFZp686P2031
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:PASK (AAH63585, 1 a.a. ~ 100 a.a) partial recombinant protein with GST tag.MEDGGLTAFEEDQRCLSQSLPLPVSAEGPAAQTTAEPSRSFSSAHRHLSRRNGLSRLCQSRTALSEDRWSSYCLSSLAAQNICTSKLHCPAAPEHTDPSE
- Purification:IgG purified
- Cat. No.:103341-056
- Supplier no.:H00023178-M01
Specifications
About this item
The PASK Antibody (6D10) from Novus Biologicals is a mouse monoclonal antibody to PASK. This antibody reacts with human. The PASK Antibody (6D10) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: PASK
Clonality: Monoclonal
Clone: 6D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human