Anti-OXCT1 Rabbit Polyclonal Antibody
Catalog # 103332-154
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:OXCT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μg
- Gene ID:5019
- Antigen Synonyms:EC 2.8.3|SCOTmitochondrial|EC 2.8.3.5|3-oxoacid CoA transferase 1,3-oxoacid CoA transferase
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:OXCT1 (NP_000427.1, 1 a.a. - 520 a.a.) full-length human protein. MAALKLLSSGLRLCASARGSGATWYKGCVCSFSTSAHRHTKFYTDPVEAVKDIPDGATVLVGGFGLCGIPENLIDALLKTGVKGLTAVSNNAGVDNFGLGLLLRSKQIKRMVSSYVGENAEFERQYLSGELEVELTPQGTLAERIRAGGAGVPAFYTPTGYGTLVQEGGSPIKYNKDGSVAIASKPREVREFNGQHFILEEAITGDFALVKAWKADRAGNVIFRKSARNFNLPMCKAAETTVVEVEEIVDIGAFAPEDIHIPQIYVHRLIKGEKYEKRIERLSIRKEGDGEAKSAKPGDDVRERIIKRAALEFEDGMYANLGIGIPLLASNFISPNITVHLQSENGVLGLGPYPRQHEADADLINAGKETVTILPGASFFSSDESFAMIRGGHVDLTMLGAMQVSKYGDLANWMIPGKMVKGMGGAMDLVSSAKTKVVVTMEHSAKGNAHKIMEKCTLPLTGKQCVNRIITEKAVFDVDKKKGLTLIELWEGLTVDDVQKSTGCDFAVSPKLMPMQQIAN
- Purification:Protein G purified
- Cat. No.:103332-154
- Supplier no.:H00005019-D01P
Specifications
About this item
The OXCT1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to OXCT1. This antibody reacts with human, mouse. The OXCT1 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: OXCT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse