Anti-ODCEC Mouse Monoclonal Antibody [clone: 2G5]
103332-064
- Antibody Type:Primary
- Antigen Name:Ornithine Decarboxylase
- Antigen Symbol:ODCEC
- Clonality:Monoclonal
- Clone:2G5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b kappa
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:100 μg
- Gene ID:4953
- Antigen Synonyms:ornithine decarboxylase|ODCEC 4.1.1.17|ornithine decarboxylase 1
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:ODC1 (AAH25296, 1 a.a. - 461 a.a.) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MNNFGNEEFDCHFLDEGFTAKDILDQKINEVSSSDDKDAFYVADLGDILKKHLRWLKALPRVTPFYAVKCNDSKAIVKTLAATGAGFDCASKTEIQLVQSLGVPPERIIYANPCKQVSQIKYAANNGVQMMTFDSEVELMKVARAHPKAKLVLRIATDDSKAVCRLSVKFGATLRTSRLLLERAKELNIDVVGVSFHVGSGCTDPETFVQAISDARCVFDMGAEVGFSMYLLDIGGGFPGSEDVKLKFEEITGVINPALDKYFPSDSGVRIIAEPGRYYVASAFTLAVNIIAKKIVLKEQTGSDDEDESSEQTFMYYVNDGVYGSFNCILYDHAHVKPLLQKRPKPDEKYYSSSIWGPTCDGLDRIVERCDLPEMHVGDWMLFENMGAYTVAAASTFNGFQRPTIYYVMSGPAWQLMQQFQNPDFPPEVEEQDASTLPVSCAWESGMKRHRAACASASINV
- Purification:IgG purified
- Cat. No.:103332-064
- Supplier no.:H00004953-M01
The Ornithine Decarboxylase Antibody (2G5) from Novus Biologicals is a mouse monoclonal antibody to Ornithine Decarboxylase. This antibody reacts with human, rat. The Ornithine Decarboxylase Antibody (2G5) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence, Immunoprecipitation.
Type: Primary
Antigen: ODCEC
Clonality: Monoclonal
Clone: 2G5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Rat