Anti-NSUN4 Rabbit Polyclonal Antibody
Catalog # 103258-844
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:NSUN4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:387338
- Antigen Synonyms:EC 2.1.1.-|NOL1/NOP2/Sun domain family 4|member 4|EC 2.1.1.29|EC 2.1.1|putative methyltransferase NSUN4|NOL1/NOP2/Sun domain family|MGC22960|NOP2/Sun domain family|NOL1/NOP2/Sun domain family member 4|RP4-603I14.2
- Storage Buffer:PBS (pH 7.2) and 40% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed against Recombinant Protein corresponding to amino acids:MAALTLRGVRELLKRVDLATVPRRHRYKKKWAATEPKFPAVRLALQNFDMTYSVQFGDLWPSIRVSLLSEQKYGALVNNFAAWDHVSAKLEQLSAKD
- Purification:Immunogen affinity purified
- Cat. No.:103258-844
- Supplier no.:NBP1-82733
Specifications
About this item
The NSUN4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to NSUN4. This antibody reacts with human. The NSUN4 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: NSUN4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human