To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:NLRP3/NALP3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:114548
- Antigen Synonyms:MWS|AII|Muckle-Wells syndrome|LRR and PYD domains-containing protein 3|FCU|NLRP3|NACHT|and PYD-containing protein 3|NALP3|AII/AVP|C1orf7|CLR1.1|AVP|Cold autoinflammatory syndrome 1|Leucine Rich Repeat And Pyrin Domain Containing 3|Nucleotide-Binding Oligomerization Domain|Angiotensin/vasopressin receptor AII/AVP-like|Leucine-rich repeat-|NLR family|PYRIN-containing APAF1-like protein 1|pyrin domain containing 3|Cryopyrin|PYPAF1|Caterpiller protein 1.1|Familial cold autoinflammatory syndrome|Cold autoinflammatory syndrome 1 protein|LRR and PYD containing protein 3
- Storage Buffer:PBS (pH 7.2) and 40% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed against Recombinant Protein corresponding to amino acids:FKMHLEDYPPQKGCIPLPRGQTEKADHVDLATLMIDFNGEEKAWAMAVWIFAAINRRDLYEKAKRDEPKWGSDNARVSNPTVICQEDSIEEEWMGLLEYLSRISICKMKKDYRKKYR
- Purification:Immunogen affinity purified
- Cat. No.:103280-178
- Supplier no.:NBP1-90207
Specifications
About this item
The NLRP3 / NALP3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to NLRP3 / NALP3. This antibody reacts with human. The NLRP3 / NALP3 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: NLRP3/NALP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human