Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Symbol:NKG2C/CD159c/KLRC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:3822
- Antigen Synonyms:NKG2-C|killer cell lectin-like receptor subfamily C|MGC138244|CD159 antigen-like family member C|CD159c antigen|NKG2-C type II integral membrane protein|member 2|NK cell receptor C|NKG2CNKG2-C-activating NK receptor|CD159c
- Storage Buffer:0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Concentration:0.56 mg/ml
- Immunogen:Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human NKG2C. The exact sequence is proprietary.
- Purification:Immunogen affinity purified
- Cat. No.:102709-106
- Supplier no.:NBP2-17085
Specifications
About this item
The NKG2C / CD159c / KLRC2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to NKG2C / CD159c / KLRC2. This antibody reacts with human. The NKG2C / CD159c / KLRC2 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: NKG2C/CD159c/KLRC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human