Anti-NHE6/SLC9A6 Mouse Monoclonal Antibody [clone: 2D5]
Catalog # 103338-992
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:NHE6/SLC9A6
- Clonality:Monoclonal
- Clone:2D5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:10479
- Antigen Synonyms:member 6|Solute carrier family 9 member 6|solute carrier family 9 (sodium/hydrogen exchanger)|NHE6isoform 6|MRSA
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:SLC9A6 (NP_006350, 602 a.a. ~ 670 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.YGDSTVNTEPATSSAPRRFMGNSSEDALDRELAFGDHELVIRGTRLVLPMDDSEPPLNLLDNTRHGPA
- Purification:IgG purified
- Cat. No.:103338-992
- Supplier no.:H00010479-M02
Specifications
About this item
The NHE6 / SLC9A6 Antibody (2D5) from Novus Biologicals is a mouse monoclonal antibody to NHE6 / SLC9A6. This antibody reacts with human. The NHE6 / SLC9A6 Antibody (2D5) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: NHE6/SLC9A6
Clonality: Monoclonal
Clone: 2D5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human