Anti-NG2/MCSP Mouse Monoclonal Antibody (DyLight 405) [clone: LHM 2]
Catalog # 103357-324
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:NG2/MCSP
- Clonality:Monoclonal
- Clone:LHM 2
- Conjugation:DyLight® 405
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:1464
- Antigen Synonyms:chondroitin sulfate proteoglycan 4 (melanoma-associated)|MCSPChondroitin sulfate proteoglycan NG2|MSK16Melanoma-associated chondroitin sulfate proteoglycan|EC 3.6.3|MEL-CSPG|MCSPGMelanoma chondroitin sulfate proteoglycan|NG2EC 2.7.8|chondroitin sulfate proteoglycan 4|HMW-MAA
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:A 375P cells crude extract
- Purification:Protein G purified
- Cat. No.:103357-324
Specifications
About this item
The NG2 / MCSP Antibody (LHM 2) [DyLight 405] from Novus Biologicals is a mouse monoclonal antibody to NG2 / MCSP. This antibody reacts with human. The NG2 / MCSP Antibody (LHM 2) [DyLight 405] has been validated for the following applications: Western Blot, Flow Cytometry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Type: Primary
Antigen: NG2/MCSP
Clonality: Monoclonal
Clone: LHM 2
Conjugation: DyLight 405
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human