Anti-NELL1 Mouse Monoclonal Antibody [clone: 6A8]
Catalog # 103331-596
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:NELL1
- Clonality:Monoclonal
- Clone:6A8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:4745
- Antigen Synonyms:FLJ45906|NRP1|nel (chicken)-like 1|neural epidermal growth factor-like 1|IDH3GL|NEL-like protein 1|NEL-like 1 (chicken)|Nel-related protein 1|protein kinase C-binding protein NELL1
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:NELL1 (NP_006148 304 a.a. - 403 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. CRRMSCPPLNCSPDSLPVHIAGQCCKVCRPKCIYGGKVLAEGQRILTKSCRECRGGVLVKITEMCPPLNCSEKDHILPENQCCRVCRGHNFCAEGPKCGE
- Purification:IgG purified
- Cat. No.:103331-596
- Supplier no.:H00004745-M01
Specifications
About this item
The NELL1 Antibody (6A8) from Novus Biologicals is a mouse monoclonal antibody to NELL1. This antibody reacts with human. The NELL1 Antibody (6A8) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence, Immunoprecipitation.
Type: Primary
Antigen: NELL1
Clonality: Monoclonal
Clone: 6A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human