- Description:Labstix Urinalysis Reagent Test Strips
- Cat. No.:10789-364
- Parameters tested:Blood, Ketone, Glucose, Protein, pH
- Supplier no.:2181
Labstix® Urinalysis Reagent Test Strips with ID Bands from Siemens Healthcare Diagnostics trigger important automatic checks (Auto-Checks) when used with CLINITEK® Analyzers.
- Auto-Check feature
- Saves time
- Reduces possible errors
Minimizes false-positive results with automatic notification if test strip has been degraded from humidity, preventing potentially erroneous patient results from being reported
The new test strips can be used on Siemens’ CLINITEK® 50, CLINITEK® 100, CLINITEK Status® (software versions 1.900 and earlier), CLINITEK® 500 and CLINITEK Advantus® analyzers version 2.0 or earlier, without impacting performance. However, since these analyzers do not recognize the ID bands, the Auto-Checks features are not available. CLINITEK Advantus® Analyzers with versions 3.0 or greater will have Auto-Checks enabled.
Enables user compliance with IVDD regulations by reporting results only from urinalysis strips that have been validated for use on the CLINITEK Status analyzers. Provides printed sample interference notes when common sample interferences are detected, providing healthcare professionals additional clinical information to aid in patient management. Eliminates the need to manually select strip type if using different urinalysis strips. Saves time and reduces possible errors.