Orders LinkContactUsLinkComponent
Albustix® Urinalysis Reagent Test Strips, Siemens Healthineers
Albustix® Urinalysis Reagent Test Strips, Siemens Healthineers
Albustix Urinalysis Reagent Test Strips, Protein
Catalog # 10789-366
Supplier #:  10333485
Albustix® Urinalysis Reagent Test Strips, Siemens Healthineers
Catalog # 10789-366
CAS Number:  
Albustix® Urinalysis Reagent Test Strips, Siemens Healthineers
Albustix® Urinalysis Reagent Test Strips, Siemens Healthineers
Albustix Urinalysis Reagent Test Strips, Protein
CAS Number:  
Price
$62.83
Log in for availability
Each Case of 12
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Albustix® Urinalysis Reagent Test Strips from Siemens Healthineers are a dip-and-read test for protein in urine.
  • Dip-and-read test
  • Detects protein in urine
Each bottle contains 100 strips.
Specifications
Specifications
  • Description:
    Albustix Urinalysis Reagent Test Strips
  • Parameters tested:
    Protein