Anti-Nectin-3/PVRL3 Mouse Monoclonal Antibody [clone: 1D1]
103341-974
- Antibody Type:Primary
- Antigen Name:Nectin-3/PVRL3
- Clonality:Monoclonal
- Clone:1D1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:25945
- Antigen Synonyms:nectin-3|PPR3|nectin 3|PRR3CD113|poliovirus receptor-related protein 3|poliovirus receptor-related 3|PVRR3|DKFZp566B0846|CDw113FLJ90624|CD113 antigen
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:PVRL3 (NP_056295, 59 a.a. ~ 167 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
- Purification:IgG purified
- Cat. No.:103341-974
- Supplier no.:H00025945-M01
The Nectin-3 / PVRL3 Antibody (1D1) from Novus Biologicals is a mouse monoclonal antibody to Nectin-3 / PVRL3. This antibody reacts with human. The Nectin-3 / PVRL3 Antibody (1D1) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: Nectin-3/PVRL3
Clonality: Monoclonal
Clone: 1D1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human