Anti-Lunatic Fringe Mouse Monoclonal Antibody [clone: 2D10-3C11]
Catalog # 103330-136
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Name:Lunatic Fringe
- Clonality:Monoclonal
- Clone:2D10-3C11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:3955
- Antigen Synonyms:O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase|beta-1,3-N-acetylglucosaminyltransferase lunatic fringe|SCDO3EC 2.4.1.222|lunatic fringe (Drosophila) homolog|lunatic fringe homolog (Drosophila)|LFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:LFNG (AAH14851, 1 a.a. - 250 a.a.) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MTPGRCCLAADIQVETFIFTDGEDEALARHTGNVVITNCSAAHSRQALSCKMAVEYDRFIESGRKWFCHVDDDNYVNLRALLRLLASYPHTRDVYVGKPSLDRPIQAMERVSENKVRPVHFWFATGGAGFCISRGLALKMSPWASGGHFMNTAERIRLPDDCTIGYIVEALLGVPLIRSGLFHSHLENLQQVPTSELHEQVTLSYGMFENKRNAVHVKGPFSVEADPSRFRSIHCHLYPDTPWCPRTAIF
- Purification:IgG purified
- Cat. No.:103330-136
- Supplier no.:H00003955-M01
Specifications
About this item
The Lunatic Fringe Antibody (2D10-3C11) from Novus Biologicals is a mouse monoclonal antibody to Lunatic Fringe. This antibody reacts with human. The Lunatic Fringe Antibody (2D10-3C11) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: Lunatic Fringe
Clonality: Monoclonal
Clone: 2D10-3C11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human