Anti-Keratin 36 Mouse Polyclonal Antibody
103335-430
- Antibody Type:Primary
- Antigen Name:Keratin 36
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Gene ID:8689
- Antigen Synonyms:keratin-36|K36|6|hair|HKA6|type I Ha6|KRTHA6keratin|HA6|keratin 36|type I cuticular Ha6|acidic|Hair keratin|HHA6|keratin
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:KRT36 (AAH43581.1, 1 a.a. - 417 a.a.) full-length human protein. MLRSGLSGLGSCLPGSYLSSECHTSGFVGSGGWFCEGSFNGSEKETMQFLNDRLANYLEKVRQLERENAELESRIQEWYEFQIPYICPDYQSYFKTIEDFQQKILLTKSENARLVLQIDNAKLAADDFRTKYETELSLRQLVEADINGLRRILDELTLCKADLEAQVESLKEELMCLKKNHEEEVSVLRCQLGDRLNVEVDAAPPVDLNKILEDMRCQYEALVENNRRDVEAWFNTQTEELNQQVVSSSEQLQCCQTEIIELRRTVNALEIELQAQHSMRNSLESTLAETEARYSSQLAQMQCLISNVEAQLSEIRCDLERQNQEYQVLLDVKARLEGEIATYRHLLEGEDCKLPPQPCATACKPVIRVPSVPPVPCVPSVPCTPAPQVGTQIRTITEEIRDGKVISSREHVQSRPL
- Purification:Protein A purified
- Cat. No.:103335-430
- Supplier no.:H00008689-B01P
The Keratin 36 Antibody from Novus Biologicals is a mouse polyclonal antibody to Keratin 36. This antibody reacts with human. The Keratin 36 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: Keratin 36
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human