Anti-Kappa Light Chain Mouse Monoclonal Antibody [clone: HP6053 + L1C1]
103281-954
- Antibody Type:Primary
- Antigen Name:Kappa Light Chain
- Clonality:Monoclonal
- Clone:HP6053 + L1C1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:7 ml
- Gene ID:3514
- Antigen Synonyms:immunoglobulin kappa constant|Km|MGC88770|MGC88771|MGC72072|MGC88809|IGKCD|HCAK1|MGC62011|MGC111575
- Storage Buffer:10mM PBS and 0.05% BSA
- Storage Temperature:Store at 4C.
- Immunogen:Purified human Ig kappa chain (HP6053) & Human B-Lymphoma Cells (L1C1)
- Purification:Protein A or G purified
- Cat. No.:103281-954
- Supplier no.:NBP2-45085
The Kappa Light Chain Antibody (HP6053 + L1C1) - IHC-Prediluted from Novus Biologicals is a mouse monoclonal antibody to Kappa Light Chain. This antibody reacts with human. The Kappa Light Chain Antibody (HP6053 + L1C1) - IHC-Prediluted has been validated for the following applications: Immunohistochemistry-Paraffin.
Type: Primary
Antigen: Kappa Light Chain
Clonality: Monoclonal
Clone: HP6053 + L1C1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human