Anti-Involucrin Mouse Monoclonal Antibody [clone: SPM259]
Catalog # 103288-752
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Name:Involucrin
- Clonality:Monoclonal
- Clone:SPM259
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Primates,Pig,Human,Dog
- Size:7 ml
- Gene ID:3713
- Antigen Synonyms:involucrin
- Storage Buffer:10mM PBS and 0.05% BSA
- Storage Temperature:Store at 4C.
- Immunogen:Purified involucrin from human keratinocytes
- Purification:Protein A or G purified
- Cat. No.:103288-752
- Supplier no.:NBP2-44882
Specifications
About this item
The Involucrin Antibody (SPM259) - IHC-Prediluted from Novus Biologicals is a mouse monoclonal antibody to Involucrin. This antibody reacts with human, porcine, canine, primate, mouse (negative). The Involucrin Antibody (SPM259) - IHC-Prediluted has been validated for the following applications: Immunohistochemistry-Paraffin.
Type: Primary
Antigen: Involucrin
Clonality: Monoclonal
Clone: SPM259
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Porcine, Canine, Primate, Mouse (Negative)