Anti-Glycogen phosphorylase, muscle form Mouse Monoclonal Antibody [clone: 2C4]
Catalog # 103308-850
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Name:Glycogen phosphorylase, muscle form
- Clonality:Monoclonal
- Clone:2C4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Gene ID:5837
- Antigen Synonyms:muscle form|muscle|glycogen phosphorylase|glycogen|EC 2.4.1.1|myophosphorylase|phosphorylase
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:PYGM (NP_005600.1 734 a.a. - 842 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. DRIPELRQVIEQLSSGFFSPKQPDLFKDIVNMLMHHDRFKVFADYEDYIKCQEKVSALYKNPREWTRMVIRNIATSGKFSSDRTIAQYAREIWGVEPSRQRLPAPDEAI
- Purification:Protein A purified
- Cat. No.:103308-850
- Supplier no.:H00005837-M10
Specifications
About this item
The Glycogen phosphorylase, muscle form Antibody (2C4) from Novus Biologicals is a mouse monoclonal antibody to Glycogen phosphorylase, muscle form. This antibody reacts with human, mouse, rat. The Glycogen phosphorylase, muscle form Antibody (2C4) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: Glycogen phosphorylase, muscle form
Clonality: Monoclonal
Clone: 2C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat