Order Entry
United States
ContactUsLinkComponent
 
Anti-NEDD9/CASL/HEF1 Rabbit Polyclonal Antibody
  103331-588
 :  Avantor
Anti-NEDD9/CASL/HEF1 Rabbit Polyclonal Antibody
  103331-588
 :  Avantor

 

  • Antibody Type:
    Primary
  • Antigen Symbol:
    NEDD9/CASL/HEF1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 μg
  • Gene ID:
    4739
  • Antigen Synonyms:
    CAS2|developmentally down-regulated 9|enhancer of filamentation 1|CASLdJ49G10.2|Cas scaffolding protein family member 2|hEF1|Neural precursor cell expressed developmentally down-regulated protein 9|dJ49G10.2 (Enhancer of Filamentation 1 (HEF1))|dJ761I2.1|neural precursor cell expressed|dJ761I2.1 (enhancer of filamentation (HEF1))|CRK-associated substrate-related protein|Renal carcinoma antigen NY-REN-12|HEF1Crk-associated substrate related|NEDD-9|p105|CasL|CAS-LCASS2cas-like docking
  • Storage Buffer:
    PBS (pH 7.4)
  • Storage Temperature:
    Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
  • Immunogen:
    NEDD9 (NP_006394.1, 1 a.a. - 834 a.a.) full-length human protein. MKYKNLMARALYDNVPECAEELAFRKGDILTVIEQNTGGLEGWWLCSLHGRQGIVPGNRVKLLIGPMQETASSHEQPASGLMQQTFGQQKLYQVPNPQAAPRDTIYQVPPSYQNQGIYQVPTGHGTQEQEVYQVPPSVQRSIGGTSGPHVGKKVITPVRTGHGYVYEYPSRYQKDVYDIPPSHTTQGVYDIPPSSAKGPVFSVPVGEIKPQGVYDIPPTKGVYAIPPSACRDEAGLREKDYDFPPPMRQAGRPDLRPEGVYDIPPTCTKPAGKDLHVKYNCDIPGAAEPVARRHQSLSPNHPPPQLGQSVGSQNDAYDVPRGVQFLEPPAETSEKANPQERDGVYDVPLHNPPDAKGSRDLVDGINRLSFSSTGSTRSNMSTSSTSSKESSLSASPAQDKRLFLDPDTAIERLQRLQQALEMGVSSLMALVTTDWRCYGYMERHINEIRTAVDKVELFLKEYLHFVKGAVANAACLPELILHNKMKRELQRVEDSHQILSQTSHDLNECSWSLNILAINKPQNKCDDLDRFVMVAKTVPDDAKQLTTTINTNAEALFRPGPGSLHLKNGPESIMNSTEYPHGGSQGQLLHPGDHKAQAHNKALPPGLSKEQAPDCSSSDGSERSWMDDYDYVHLQGKEEFERQQKELLEKENIMKQNKMQLEHHQLSQFQLLEQEITKPVENDISKWKPSQSLPTTNSGVSAQDRQLLCFYYDQCETHFISLLNAIDALFSCVSSAQPPRIFVAHSKFVILSAHKLVFIGDTLTRQVTAQDIRNKVMNSSNQLCEQLKTIVMATKMAALHYPSTTALQEMVHQVTDLSRNAQLFKRSLLEMATF
  • Purification:
    Protein A purified
  • Cat. No.:
    103331-588
  • Supplier no.:
    H00004739-D01P

 

 

The NEDD9 / CASL / HEF1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to NEDD9 / CASL / HEF1. This antibody reacts with human, mouse. The NEDD9 / CASL / HEF1 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: NEDD9/CASL/HEF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse