Anti-Ndufs4 Mouse Monoclonal Antibody [clone: 3F11]
Catalog # 103331-564
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:Ndufs4
- Clonality:Monoclonal
- Clone:3F11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:4724
- Antigen Synonyms:NADH-coenzyme Q reductase|18kDa (NADH-coenzyme Qreductase)|NADH dehydrogenase (ubiquinone) Fe-S protein 4|NADH dehydrogenase (ubiquinone) Fe-S protein 4 (18kD) (NADH-coenzyme Qreductase)|AQDQmitochondrial|AQDQ|18-KD|NADH-ubiquinone oxidoreductase 18 kDa subunit|CI-18 kDa
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:NDUFS4 (NP_002486, 66 a.a. - 175 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. GVPEEHIKTRKVRIFVPARNNMQSGVNNTKKWKMEFDTRERWENPLMGWASTADPLSNMVLTFSTKEDAVSFAEKNGWSYDIEERKVPKPKSKSYGANFSWNKRTRVSTK
- Purification:IgG purified
- Cat. No.:103331-564
- Supplier no.:H00004724-M03
Specifications
About this item
The Ndufs4 Antibody (3F11) from Novus Biologicals is a mouse monoclonal antibody to Ndufs4. This antibody reacts with human. The Ndufs4 Antibody (3F11) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: Ndufs4
Clonality: Monoclonal
Clone: 3F11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human