Anti-NDUFB3 Rabbit Polyclonal Antibody
Catalog # 103331-528
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:NDUFB3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:4709
- Antigen Synonyms:NADH dehydrogenase (ubiquinone) 1 beta subcomplex|B12)|12kDa|3 (12kD|3
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:NDUFB3 (NP_002482.1, 1 a.a. - 98 a.a.) full-length human protein. MAHEHGHEHGHHKMELPDYRQWKIEGTPLETIQKKLAAKGLRDPWGRNEAWRYMGGFAKSVSFSDVFFKGFKWGFAAFVVAVGAEYYLESLNKDKKHH
- Purification:Protein A purified
- Cat. No.:103331-528
- Supplier no.:H00004709-D01P
Specifications
About this item
The NDUFB3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to NDUFB3. This antibody reacts with human. The NDUFB3 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: NDUFB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human