Anti-MYLIP Mouse Monoclonal Antibody [clone: 3C10]
103343-016
- Antibody Type:Primary
- Antigen Symbol:MYLIP
- Clonality:Monoclonal
- Clone:3C10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Size:100 μg
- Gene ID:29116
- Antigen Synonyms:EC 6.3.2.-|MIRE3 ubiquitin-protein ligase MYLIP|Idol|BZF1|band 4.1 superfamily member BZF1|Inducible degrader of the LDL-receptor|IDOLmyosin regulatory light chain-interacting protein|cellular modulator of immune recognition (c-MIR)|myosin regulatory light chain interacting proteinE3 ubiquitin ligase-inducible degrader of the low density lipoprotein receptor
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:MYLIP (AAH02860, 1 a.a. ~ 446 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.MLCYVTRPDAVLMEVEVEAKANGEDCLNQVCRRLGIIEVDYFGLQFTGSKGESLWLNLRNRISQQMDGLAPYRLKLRVKFFVEPHLILQEQTRHIFFLHIKEALLAGHLLCSPEQAVELSALLAQTKFGDYNQNTAKYNYEELCAKELSSATLNSIVAKHKELEGTSQASAEYQVLQIVSAMENYGIEWHSVRDSEGQKLLIGVGPEGISICKDDFSPINRIAYPVVQMATQSGKNVYLTVTKESGNSIVLLFKM
- Purification:IgG purified
- Cat. No.:103343-016
- Supplier no.:H00029116-M02
The MYLIP Antibody (3C10) from Novus Biologicals is a mouse monoclonal antibody to MYLIP. This antibody reacts with human. The MYLIP Antibody (3C10) has been validated for the following applications: ELISA.
Type: Primary
Antigen: MYLIP
Clonality: Monoclonal
Clone: 3C10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human