Anti-MTH1 Rabbit Polyclonal Antibody (Alexa Fluor® 700)
Catalog # 103367-910
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:MTH1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 700
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:0.2 ml
- Gene ID:4521
- Antigen Synonyms:EC 3.6.1.-|8-oxo-7,8-dihydrodeoxyguanosine triphosphatase|mutT human homolog 1,8-oxo-dGTPase|nudix (nucleoside diphosphate linked moiety X)-type motif 1,8-oxo-7,8-dihydroguanosine triphosphatase|MTH1Nucleoside diphosphate-linked moiety X motif 1|Nudix motif 1|nucleoside diphosphate-linked moiety X-type motif 17,8-dihydro-8-oxoguanine triphosphatase
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:A C-terminal peptide derived from human MTH1, conjugated to KLH. [UniProt# P36639]
- Purification:Immunogen affinity purified
- Cat. No.:103367-910
Specifications
About this item
The MTH1 Antibody [Alexa Fluor« 700] from Novus Biologicals is a rabbit polyclonal antibody to MTH1. This antibody reacts with human, mouse, rat. The MTH1 Antibody [Alexa Fluor« 700] has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Type: Primary
Antigen: MTH1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 700
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat