Anti-MTG1 Mouse Polyclonal Antibody
Catalog # 103345-866
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:MTG1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Gene ID:92170
- Antigen Synonyms:mitochondrial GTPase 1 homolog (S. cerevisiae)|GTPBP7|RP11-108K14.2|GTP-binding protein 7|mitochondrial GTPase 1|GTP-binding protein 7 (putative)|GTP
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:MTG1 (AAH35721.1, 1 a.a. ~ 300 a.a) full-length human protein.MAKGLKKMQSSLKLVDCIIEVHDARIPLSGRNPLFQETLGLKPHLLVLNKMDLADLTEQQKIMQHLEGEGLKNVIFTNCVKDENVKQIIPMVTELIGRSHRYHRKENLEYCIMVIGVPNVGKSSLINSLRRQHLRKGKATRVGGEPGITRAVMSKIQVSERPLMFLLDTPGVLAPRIESVETGLKLALCGTVLDHLVGEETMADYLLYTLNKHQRFGYVQHYGLGSACDNVERVLKSVAVKLGKTQKVKVLTGTG
- Purification:Protein A purified
- Cat. No.:103345-866
- Supplier no.:H00092170-B01P
Specifications
About this item
The MTG1 Antibody from Novus Biologicals is a mouse polyclonal antibody to MTG1. This antibody reacts with human. The MTG1 Antibody has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: MTG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human