To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:MRP6
- Clonality:Monoclonal
- Clone:5D8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:368
- Antigen Synonyms:pseudoxanthoma elasticum|EST349056|MOAT-E|Anthracycline resistance-associated protein|member 6|Multi-specific organic anion transporter E|MOATE|sub-family C (CFTR/MRP)|multidrug resistance-associated protein 6|MRP6ATP-binding cassette sub-family C member 6|ATP-binding cassette|URG7|ARAPXE|MLP1ABC34|PXE1
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:ABCC6 (NP_001162, 831 a.a. - 930 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. IAEMGSYQELLQRKGALVCLLDQARQPGDRGEGETEPGTSTKDPRGTSAGRRPELRRERSIKSVPEKDRTTSEAQTEVPLDDPDRAGWPAGKDSIQYGRV
- Purification:IgG purified
- Cat. No.:103322-032
- Supplier no.:H00000368-M02
Specifications
About this item
The MRP6 Antibody (5D8) from Novus Biologicals is a mouse monoclonal antibody to MRP6. This antibody reacts with human. The MRP6 Antibody (5D8) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: MRP6
Clonality: Monoclonal
Clone: 5D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human