Anti-Motilin Mouse Monoclonal Antibody [clone: 4E12.]
Catalog # 103330-924
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Name:Motilin
- Antigen Symbol:Motilin
- Clonality:Monoclonal
- Clone:4E12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Cross Adsorption:No
- Gene ID:4295
- Antigen Synonyms:prepromotilin|MGC138519|motilin|promotilin
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:MLN (AAH69675.1, 1 a.a. - 115 a.a.) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MVSRKAVAALLVVHAAAMLASQTEAFVPIFTYGELQRMQEKERNKGQKKSLSVWQRSGEEGPVDPAEPIREEENEMIKLTAPLEIGMRMNSRQLEKYPATLEGLLSEMLPQHAAK
- Purification:IgG purified
- Cat. No.:103330-924
- Supplier no.:H00004295-M07
Specifications
About this item
The Motilin Antibody (4E12.) from Novus Biologicals is a mouse monoclonal antibody to Motilin. This antibody reacts with human. The Motilin Antibody (4E12.) has been validated for the following applications: Western Blot, ELISA.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: Motilin
Clonality: Monoclonal
Clone: 4E12.
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human