Anti-MGRN1 Rabbit Polyclonal Antibody
Catalog # 103266-378
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:MGRN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 μl
- Gene ID:23295
- Antigen Synonyms:KIAA0544probable E3 ubiquitin-protein ligase MGRN1|E3 ubiquitin-protein ligase MGRN1|ring finger 1|Mahogunin RING finger protein 1|mahogunin|RNF156EC 6.3.2.-|RING finger protein 156|EC 6.3.2
- Storage Buffer:PBS (pH 7.2) and 40% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed against Recombinant Protein corresponding to amino acids:LPFRALLQIRAVRKKPGALSPVSFSPVLAQSLEHDEHSNSDSVPPGYEPISLLEALNGLRAVSPAIPSAPLYEEITYSGISDGLSQASCPLAAIDHILDSSRQKGRPQSKAPDSTLRSPSSPIHEEDE
- Purification:Immunogen affinity purified
- Cat. No.:103266-378
- Supplier no.:NBP1-82998
Specifications
About this item
The MGRN1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to MGRN1. This antibody reacts with human. The MGRN1 Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: MGRN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human